Anti-schistosomal Glutathione-S-Transferase Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Columbia Biosciences
AP-1310
COBSAP-1310EA 629 EUR
COBSAP-1310
Anti-schistosomal Glutathione-S-Transferase Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type: Primary
Antigen: schistosomal Glutathione-S-Transferase
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR