Anti-HA tag Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 16B12]

Lieferant: Columbia Biosciences
AP-1722
COBSAP-1722EA 629 EUR
COBSAP-1722
Anti-HA tag Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 16B12]
Antikörper
Type: Tag
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone: 16B12
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR