Anti-V5 Tag Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Columbia Biosciences
AP-1834
COBSAP-1834EA 629 EUR
COBSAP-1834
Anti-V5 Tag Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Type: Tag
Antigen: V5 Tag (V5 tag)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR