Anti-6X His tag Mouse Monoclonal Antibody (Biotin) [clone: AD1.1.10]

Lieferant: Columbia Biosciences
B1-1711
COBSB1-1711EA 629 EUR
COBSB1-1711
Anti-6X His tag Mouse Monoclonal Antibody (Biotin) [clone: AD1.1.10]
Antikörper
Type: Tag
Antigen: 6X His tag (6X His tag)
Clonality: monoclonal
Clone: AD1.1.10
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR