Anti-c-Myc tag Mouse Monoclonal Antibody (Biotin) [clone: 9E10]
Lieferant: Columbia Biosciences
B1-1714
COBSB1-1714EA 629 EUR
COBSB1-1714
Anti-c-Myc tag Mouse Monoclonal Antibody (Biotin) [clone: 9E10]
Antikörper
Type: Tag
Antigen: c-Myc tag (c-Myc tag)
Clonality: monoclonal
Clone: 90000000000
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen