Anti-S Tag Rabbit Polyclonal Antibody (DyLight® 650)
Lieferant: Columbia Biosciences
D10-1831
COBSD10-1831EA 629 EUR
COBSD10-1831
Anti-S Tag Rabbit Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Tag
Antigen: S Tag (S tag)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen