Anti-V5 Tag Rabbit Polyclonal Antibody (DyLight® 650)

Lieferant: Columbia Biosciences
D10-1834
COBSD10-1834EA 629 EUR
COBSD10-1834
Anti-V5 Tag Rabbit Polyclonal Antibody (DyLight® 650)
Antikörper
Type: Tag
Antigen: V5 Tag (V5 tag)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR