Anti-schistosomal Glutathione-S-Transferase Goat Polyclonal Antibody (Europium 1024)

Lieferant: Columbia Biosciences
D17-1310-50 D17-1310-20 D17-1310-1MG
COBSD17-1310-50EA 1370 EUR
COBSD17-1310-50 COBSD17-1310-20 COBSD17-1310-1MG
Anti-schistosomal Glutathione-S-Transferase Goat Polyclonal Antibody (Europium 1024)
Antikörper
Type: Tag
Antigen: schistosomal Glutathione-S-Transferase
Clonality: polyclonal
Clone:
Conjugation: Europium 1024
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR