Anti-c-Myc tag Mouse Monoclonal Antibody (Europium 1024) [clone: 9E10]

Lieferant: Columbia Biosciences
D17-1714-20 D17-1714-50 D17-1714-1MG
COBSD17-1714-20EA 683 EUR
COBSD17-1714-20 COBSD17-1714-50 COBSD17-1714-1MG
Anti-c-Myc tag Mouse Monoclonal Antibody (Europium 1024) [clone: 9E10]
Antikörper
Type: Tag
Antigen: c-Myc tag (c-Myc tag)
Clonality: monoclonal
Clone: 90000000000
Conjugation: Europium 1024
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR