Anti-IgE Goat Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: Columbia Biosciences
D3-110-E
COBSD3-110-EEA 629 EUR
COBSD3-110-E
Anti-IgE Goat Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR