Anti-IgM Goat Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: Columbia Biosciences
D3-110-M
COBSD3-110-MEA 385 EUR
COBSD3-110-M
Anti-IgM Goat Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human