Anti-IgG2b Goat Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: Columbia Biosciences
D3-112-2B
COBSD3-112-2BEA 385 EUR
COBSD3-112-2B
Anti-IgG2b Goat Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Type: Secondary
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR