Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: Columbia Biosciences
D3-112-FAB
COBSD3-112-FABEA 385 EUR
COBSD3-112-FAB
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse