Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: Columbia Biosciences
D3-112-FABFC
COBSD3-112-FABFCEA 385 EUR
COBSD3-112-FABFC
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR