Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: Columbia Biosciences
D3-113
COBSD3-113EA 199 EUR
COBSD3-113
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR