Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: Columbia Biosciences
D3-113-FC
COBSD3-113-FCEA 199 EUR
COBSD3-113-FC
Anti-IgG Goat Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat