Mouse IgG1 kappa Isotype Control (APC (Allophycocyanin))

Lieferant: Columbia Biosciences
D3-1601
COBSD3-1601EA 385 EUR
COBSD3-1601
Mouse IgG1 kappa Isotype Control (APC (Allophycocyanin))
Antikörper
Type: Control
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: None
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR