Mouse IgG2b Isotype Control (APC (Allophycocyanin))

Lieferant: Columbia Biosciences
D3-1602B
COBSD3-1602BEA 385 EUR
COBSD3-1602B
Mouse IgG2b Isotype Control (APC (Allophycocyanin))
Antikörper
Type: Control
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: None
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR