Anti-cytokeratins 4, 5, 6, 8, 10, 13, and 18 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: C11]

Lieferant: Columbia Biosciences
D3-1760
COBSD3-1760EA 629 EUR
COBSD3-1760
Anti-cytokeratins 4, 5, 6, 8, 10, 13, and 18 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: C11]
Antikörper
Type: Primary
Antigen: cytokeratins 4, 5, 6, 8, 10, 13, and 18
Clonality: monoclonal
Clone: C11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR