Anti-HA tag Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: Columbia Biosciences
D3-1830
COBSD3-1830EA 629 EUR
COBSD3-1830
Anti-HA tag Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Type: Tag
Antigen: HA tag (HA tag)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR