Anti-IgM Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Lieferant: Columbia Biosciences
D5-110-M-100
COBSD5-110-M-100EA 199 EUR
COBSD5-110-M-100
Anti-IgM Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antikörper
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human