Anti-IgG1 Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Lieferant: Columbia Biosciences
D5-112-1
COBSD5-112-1EA 385 EUR
COBSD5-112-1
Anti-IgG1 Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antikörper
Type: Secondary
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR