Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Lieferant: Columbia Biosciences
D5-113-FAB
COBSD5-113-FABEA 385 EUR
COBSD5-113-FAB
Anti-IgG Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antikörper
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR