Anti-Acetyl Lysine Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 7F8]

Lieferant: Columbia Biosciences
D5-1762
COBSD5-1762EA 629 EUR
COBSD5-1762
Anti-Acetyl Lysine Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 7F8]
Antikörper
Type: Primary
Antigen: Acetyl Lysine
Clonality: monoclonal
Clone: 7F8
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR