Anti-EP2 receptor Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))

Lieferant: Columbia Biosciences
D5-1863-50
COBSD5-1863-50EA 579 EUR
COBSD5-1863-50
Anti-EP2 receptor Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Antikörper
Type: Primary
Antigen: EP2 receptor
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR