Anti-EP4 receptor C-terminal Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))

Lieferant: Columbia Biosciences
D5-1866-50
COBSD5-1866-50EA 579 EUR
COBSD5-1866-50
Anti-EP4 receptor C-terminal Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Antikörper
Type: Primary
Antigen: EP4 receptor
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR