Anti-DYKDDDDK Tag Mouse Monoclonal Antibody SureLight® P3 [clone: M2]

Lieferant: Columbia Biosciences
D7-1718
COBSD7-1718EA 629 EUR
COBSD7-1718
Anti-DYKDDDDK Tag Mouse Monoclonal Antibody SureLight® P3 [clone: M2]
Antikörper
Type: Tag
Antigen: DYKDDDDK Tag (DYKDDDDK tag)
Clonality: monoclonal
Clone: M2
Conjugation: SureLight® P3
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR