Anti-c-Myc tag Mouse Monoclonal Antibody (DyLight® 488) [clone: 9E10]

Lieferant: Columbia Biosciences
D8-1714
COBSD8-1714EA 629 EUR
COBSD8-1714
Anti-c-Myc tag Mouse Monoclonal Antibody (DyLight® 488) [clone: 9E10]
Antikörper
Type: Tag
Antigen: c-Myc tag (c-Myc tag)
Clonality: monoclonal
Clone: 90000000000
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR