Anti-T7 Tag Rabbit Polyclonal Antibody (DyLight® 488)

Lieferant: Columbia Biosciences
D8-1832
COBSD8-1832EA 629 EUR
COBSD8-1832
Anti-T7 Tag Rabbit Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Tag
Antigen: T7 Tag (T7 tag)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR