Anti-schistosomal Glutathione-S-Transferase Goat Polyclonal Antibody (DyLight® 550)

Lieferant: Columbia Biosciences
D9-1310
COBSD9-1310EA 629 EUR
COBSD9-1310
Anti-schistosomal Glutathione-S-Transferase Goat Polyclonal Antibody (DyLight® 550)
Antikörper
Type: Tag
Antigen: schistosomal Glutathione-S-Transferase
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR