Anti-HA tag Mouse Monoclonal Antibody (DyLight® 550) [clone: 16B12]
Lieferant: Columbia Biosciences
D9-1722
COBSD9-1722EA 629 EUR
COBSD9-1722
Anti-HA tag Mouse Monoclonal Antibody (DyLight® 550) [clone: 16B12]
Antikörper
Type: Tag
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone: 16B12
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen