Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Columbia Biosciences
HRP-112
COBSHRP-112EA 293 EUR
COBSHRP-112
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR