Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Columbia Biosciences
HRP-114-FC
COBSHRP-114-FCEA 293 EUR
COBSHRP-114-FC
Anti-IgG Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit