Anti-COL2 Mouse Monoclonal Antibody [clone: II-4C11]
Lieferant: Merck Millipore (Calbiochem‎)
CP18-100
CP18-100EA 555 EUR
CP18-100
Anti-COL2 Mouse Monoclonal Antibody [clone: II-4C11]
Antikörper
Type: Primary
Antigen: COL2 (collagen, type II)
Clonality: monoclonal
Clone: II-4C11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen