Anti-Vinculin (V284 or VLN01) Mouse Monoclonal Antibody [clone: V284 or VLN01]

Lieferant: Merck Millipore (Oncogene)
CP74-100
CP74-100EA 566 EUR
CP74-100
Anti-Vinculin (V284 or VLN01) Mouse Monoclonal Antibody [clone: V284 or VLN01]
Antikörper
Type: Primary
Antigen: Vinculin (V284 or VLN01)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR