BrightVision+ Anti-IgG (Poly-HRP)
Lieferant: IMMUNOLOGIC
DPVR110HRP DPVR500HRP DPVR55HRP
DPVR110HRPEA 834 EUR
DPVR110HRP DPVR500HRP DPVR55HRP
BrightVision+ Anti-IgG (Poly-HRP)
Antikörper
BrightVision histostaining reagents utilise a novel controlled polymerisation technology to prepare polymeric HRP linker antibody conjugates. Comparing to conventional biotin-streptavidin based detection kits, this kit has the advantages of higher amplification power, biotin free and more consistent immunostaining outcomes on archival tissues and on difficult-to-work antibodies.
  • High-quality secondary antibody for IHC applications, HRP labelled
  • Antigen specificity: Anti-Rabbit IgG (H+L)
Biotin-free
Ready-to-use
Standards: Meets CE/IVD standards.
Bestellinformation: Packages above 110 ml contain small empty bottles so users can create smaller working volumes and prevent possible contamination of the whole lot.
Lieferumfang: Shipped with blue ice. Store between 2 to 8 °C.
Bitte beachten Irritating to eyes, respiratory systems and skin.