Kits: family pedigree cancer gene detection, Ready-to-Load™
Lieferant: EDVOTEK
115 115-C
EDVO115EA 109 EUR
EDVO115 EDVO115-C
Kits: family pedigree cancer gene detection, Ready-to-Load™
Lehrmittel Biologie-Lehrmittel Genetics Learning Activities
In this experiment, students determine a pedigree for a family thought to be carriers of a mutation in their p53 genes. This is followed by a diagnostic agarose gel analysis to diagnose the state of the p53 gene in individual family members.
  • For six lab groups
Also required: Electrophoresis tank, power supply, micropipettes (5 to 50 µl adjustable or 40 µl fixed volume) with tips, and a white light box is recommended.
Lieferumfang: Kit includes instructions, Ready-to-Load™ DNA samples, agarose powder, practice gel loading solution, electrophoresis buffer, InstaStain® DNA stain, 1 ml calibrated drop pipette, 100 ml graduated cyclinder and microtipped transfer pipettes.