Linking food science to biotechnology kit
Lieferant: EDVOTEK
S-47
EDVOS-47EA 71.5 EUR
EDVOS-47
Linking food science to biotechnology kit
Lehrmittel Biologie-Lehrmittel Biochemistry Learning Activities
Investigate how agarose gel electrophoresis unlocks the color code used by food scientists to make colorful candies.
  • NGSS-aligned with MS-PS1
  • For 10 lab groups
  • Complete in 45 minutes
  • Explore the biochemistry behind colorful candies
  • Understand the biology behind gel electrophoresis
  • Perform agarose gel electrophoresis to separate different sized dye molecules
  • Extract, identify, and size different food dyes from popular candies
Students will extract color activities from common candies and separate the dyes on agarose gel electrophoresis. A fun lab extension involves the use of candy to build a DNA model.
Bestellinformation: DNA electrophoresis, micropipettes: 5-50 µl (optional), white light box (Optional), and microwave or hot plate are not included.
Lieferumfang: Kit includes instructions, Ready-to-Load QuickStrip™ dye samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), practice gel loading solution, disposable pipets, and microcentrifuge tubes.
Verpackung: Room temperature storage.