4-Nitrophenylphosphat
Lieferant: ENZO LIFE SCIENCES
|
Gefahr
|
Synonyme:
4-Nitrophenyldihydrogenphosphat, ortho-Phosphorsäuremono(4-nitrophenyl)ester, p-Nitrophenylphosphat, ortho-Phosphorsäure(4-nitrophenyl)ester
ADI800076
ENZOADI800076EA
476
EUR
ENZOADI800076
4-Nitrophenylphosphat
4-Nitrophenylphosphat
AP substrate.
- Used in ELISA, colorimetric detection
- Solution of p-nitrophenylphosphate in buffer
Formel:
C₆H₆NO₆P Molecular Weight: 219.09 g/mol Lagertemperatur: Gefrierschrank |
CAS-Nummer:
330-13-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...