Wash buffer concentrate
Lieferant: ENZO LIFE SCIENCES
ADI801287
ENZOADI801287EA
159
EUR
ENZOADI801287
Wash buffer concentrate
Puffer
Assay-Puffer
Liquid. TRIS buffered saline containing detergents.
For ELISA.