Neutralising reagent
Lieferant: ENZO LIFE SCIENCES
ADI801475
ENZOADI801475EA
73.7
EUR
ENZOADI801475
Neutralising reagent
Puffer
Biologische Puffer
Neutralising buffer for low pH solutions and for use with cyclic nucleotide and isoprostane kits.