3,3',5,5'-Tetramethylbenzidin (TMB) Substrat

Lieferant: ENZO LIFE SCIENCES

Synonyme: TMB

ADI801805
ENZOADI801805EA 107 EUR
ENZOADI801805
3,3',5,5'-Tetramethylbenzidin (TMB) Substrat
3,3',5,5'-Tetramethylbenzidin (TMB)

HRP substrate.


  • Used in ELISA


This TMB Substrate is intended for use as a colourimetric HRP-substrate for immunoassay development. Each bottle is sufficient for use with 5×96 wells microtiter plates.

Formel: C₁₆H₂₀N₂
Lagertemperatur: Kühlschrank
MDL: MFCD00007748
CAS-Nummer: 54827-17-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR