Tween-20 solution (10%)
Lieferant: ENZO LIFE SCIENCES
ADI801929
ENZOADI801929EA
83.6
EUR
ENZOADI801929
Tween-20 solution (10%)
Puffer
Assay-Puffer
This Tween®-20 solution (10%) is specially designed to create the Blocking Buffer and Assay Buffer in ImmunoSet products. Each bottle is sufficient for use with 5×96 wells microtiter plates if following the instructions in our ImmunoSet protocol.
- Long Term Storage: 4 °C
The Tween-20 solution may be used for any other assay development needs. Additional assay development reagents are available as companion products to our ImmunoSet products.