15-deoxy-Δ12,14-PGJ2 ELISA kit
Lieferant: ENZO LIFE SCIENCES
ADI900023
ENZOADI900023EA 632 EUR
ENZOADI900023
15-deoxy-Δ12,14-PGJ2 ELISA kit
Tests ELISAs
First-to-market 15-d-PGJ2 ELISA commercially available for eicosanoid research.
For the quantitative determination of 15-deoxy-Δ12,14-PGJ2 in culture supernatants, plasma, saliva, and urine from any species.
The 15-deoxy-Δ12,14-PGJ2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. This kit saves time and is much less time consuming than GC/MS analysis. Ready-to-use liquid color-coded reagents reduce laboratory error.
Bestellinformation: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-290 (5x96 wells).