Leptin (human), ELISA kit
Lieferant: ENZO LIFE SCIENCES
ADI900028A
ENZOADI900028AEA
618
EUR
ENZOADI900028A
Leptin (human), ELISA kit
Tests
ELISAs
For the quantitative determination of human leptin in culture supernatants, plasma, and serum.
The Leptin (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 3 hours.
Bestellinformation: Contains Microtiter plate, Standard, Antibody, Conjugate, Assay buffer 17, TMB Substrate, Stop solution 2, Wash buffer concentrate.