IgG1 (mouse), ELISA kit
Lieferant: ENZO LIFE SCIENCES
ADI900109A
ENZOADI900109AEA 640 EUR
ENZOADI900109A
IgG1 (mouse), ELISA kit
Tests ELISAs
For the quantitative determination of IgG1 in culture supernatants and serum of mouse origin.
The IgG1 (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 90 minute. Absorbance is read at 450 nm. Ideal for determining the isotype and concentration of monoclonal antibodies.
Bestellinformation: Contains GxM IgG Microtiter plate, mouse IgG1 conjugate, Assay buffer 13 concentrate, Wash buffer concentrate, mouse IgG1 standard, TMB Substrate, Stop solution 2.