IgG2b (mouse), ELISA kit
Lieferant: ENZO LIFE SCIENCES
ADI900110
ENZOADI900110EA
553
EUR
ENZOADI900110
IgG2b (mouse), ELISA kit
Tests
ELISAs
For the quantitative determination of IgG2b in culture supernatants and serum of mouse origin.
The IgG2b (mouse), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 90 minutes. Ideal for determining the isotype and concentration of monoclonal antibodies.
Bestellinformation: Contains GxM IgG Microtiter plate, mouse IgG2b conjugate, Assay buffer 13 concentrate, Wash buffer concentrate, mouse IgG2b standard, TMB Substrate, Stop solution 2.