IL-1β (Human), ELISA Kit
Lieferant: ENZO LIFE SCIENCES
For the quantitative determination of IL-1β in culture supernatant, plasma, serum, and urine of human origin.
- Sensitive - quantify down to 1 pg/ml
- Efficient - only 50 µl diluted sample required saving precious materials
- Specific - no cross-reactivity with related cytokines
- Results from up to 40 samples in duplicate in just 3 h
- Ready-to-use reagents and pre-coated plates reduce errors and save your time
The IL-1β (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in less than 3 h. Absorbance is read at 450 nm. Quantify 1 pg/ml IL-1β with lot sample volume needed saving you precious sample.
Bestellinformation: Contains standard, standard diluent buffer, coated 96-well plate, conjugate, streptavidin-HRP, streptavidin-HRP diluent, wash buffer concentrate, TMB solution, stop solution, plate sealers.