Oxytocin ELISA kit
Lieferant: ENZO LIFE SCIENCES
ADI900153A0001
ENZOADI900153A0001EA 654 EUR
ENZOADI900153A0001
Oxytocin ELISA kit
Tests ELISAs
The most sensitive, specific and flexible alternative to more costly and time-consuming methods for oxytocin detection.
For the quantitative determination of oxytocin in culture supernatants, milk, plasma, and serum from any species. Cited sample types include cerebral spinal fluid, saliva, tissue and urine.
Bestellinformation: Contains GxR IgG microtiter plate, conjugate, antibody, assay buffer, wash buffer concentrate, standard, pNpp substrate, stop solution. Alternative size available: ADI-901-153A (5 x 96 wells).