Angiotensin I ELISA Kits
Lieferant: ENZO LIFE SCIENCES
ADI900203
ENZOADI900203EA 632 EUR
ENZOADI900203
Angiotensin I ELISA Kits
Tests ELISAs
The Angiotensin I ELISA Kit is a colorimetric, competitive immunoassay kit for the quantitative determination of species independent Angiotensin I in plasma and serum samples.
  • Results in under 3,5 hours
  • Specific - low cross reactivity to similar peptides
  • Non radioactive - enzymatic assay format is safe and reliable
  • Easy to use liquid colour coded reagents reduce errors
  • Ready to use reagents and pre-coated plates reduce errors and save your time
Cross reactivity: Angiotensin I (100%), Angiotensin A (0,182%), Angiotensin II (0,083%), Angiotensin (1-12) (0,025%). Unaffected by Angiotensin (1-9), Angiotensin (1-7), Angiotensin III, Angiotensin IV, LVV-hemorphin 7, Bradykinin.
Lieferumfang: Kit contains microtiter plate, conjugate, antibody, Assay Buffer 16, wash buffer concentrate, standard, TMB substrate, Stop Solution 2, and Streptavidin-HRP.